Assay Design
Gene Transcripts
- Gene Symbol
- RPT1
- Entrez Gene ID
-
853712
- Gene Name
- Gene Aliases
-
CIM5, YKL145W, YTA3
- Chromosome Location
-
Chr.XI: 174213 - 175616 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179711.1 |
NP_012777.1 |
1 - 1 |
1356 |
75 |
View Full Product Details