Assay Design
Gene Transcripts
- Gene Symbol
- APE2
- Entrez Gene ID
-
853699
- Gene Name
- Gene Aliases
-
LAP1, YKL157W, YKL158W
- Chromosome Location
-
Chr.XI: 154991 - 158232 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179723.2 |
NP_012765.3 |
1 - 2 |
275 |
102 |
View Full Product Details