Assay Design
Gene Transcripts
- Gene Symbol
- TRP3
- Entrez Gene ID
-
853669
- Gene Name
- Gene Aliases
-
YKL211C
- Chromosome Location
-
Chr.XI: 36699 - 38153 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179776.1 |
NP_012711.1 |
1 - 1 |
1351 |
89 |
View Full Product Details