Assay Design
Gene Transcripts
- Gene Symbol
- UIP5
- Entrez Gene ID
-
853918
- Gene Name
- Gene Aliases
-
YKR044W
- Chromosome Location
-
Chr.XI: 522015 - 523346 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179834.3 |
NP_012970.3 |
1 - 1 |
1251 |
101 |
View Full Product Details