Assay Design
Gene Transcripts
- Gene Symbol
- MET1
- Entrez Gene ID
-
853943
- Gene Name
- Gene Aliases
-
MET20, YKR069W
- Chromosome Location
-
Chr.XI: 571612 - 573393 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179859.3 |
NP_012995.3 |
1 - 1 |
1646 |
62 |
View Full Product Details