Assay Design
Gene Transcripts
- Gene Symbol
- ECM4
- Entrez Gene ID
-
853951
- Gene Name
- Gene Aliases
-
GTO2, YKR076W
- Chromosome Location
-
Chr.XI: 582283 - 583395 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001179866.3 |
NP_013002.3 |
1 - 1 |
851 |
88 |
View Full Product Details