Assay Design
Gene Transcripts
- Gene Symbol
- SPO75
- Entrez Gene ID
-
850655
- Gene Name
- Gene Aliases
-
YLL005C
- Chromosome Location
-
Chr.XII: 137940 - 140546 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181825.1 |
NP_013096.1 |
1 - 1 |
2449 |
93 |
View Full Product Details