Assay Design
Gene Transcripts
- Gene Symbol
- CMS1
- Entrez Gene ID
-
850689
- Gene Name
- Gene Aliases
-
YLR003C
- Chromosome Location
-
Chr.XII: 156855 - 157730 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181890.1 |
NP_013103.1 |
1 - 1 |
52 |
73 |
View Full Product Details