Assay Design
Gene Transcripts
- Gene Symbol
- ADE16
- Entrez Gene ID
-
850715
- Gene Name
- Gene Aliases
-
YLR028C
- Chromosome Location
-
Chr.XII: 199540 - 201315 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181915.1 |
NP_013128.1 |
1 - 1 |
1654 |
64 |
View Full Product Details