Assay Design
Gene Transcripts
- Gene Symbol
- OSW2
- Entrez Gene ID
-
850743
- Gene Name
- Gene Aliases
-
YLR054C
- Chromosome Location
-
Chr.XII: 248717 - 250978 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001181941.1 |
NP_013155.2 |
1 - 2 |
37 |
84 |
View Full Product Details