Assay Design
Gene Transcripts
- Gene Symbol
- DCN1
- Entrez Gene ID
-
850819
- Gene Name
- Gene Aliases
-
YLR128W
- Chromosome Location
-
Chr.XII: 398530 - 399433 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182015.1 |
NP_013229.1 |
1 - 1 |
756 |
103 |
View Full Product Details