Assay Design
Gene Transcripts
- Gene Symbol
- ATG26
- Entrez Gene ID
-
850886
- Gene Name
- Gene Aliases
-
UGT51, YLR189C
- Chromosome Location
-
Chr.XII: 530797 - 534393 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182076.1 |
NP_013290.1 |
1 - 1 |
3356 |
96 |
View Full Product Details