Assay Design
Gene Transcripts
- Gene Symbol
- HRD3
- Entrez Gene ID
-
850904
- Gene Name
- Gene Aliases
-
YLR207W
- Chromosome Location
-
Chr.XII: 556788 - 559289 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182094.1 |
NP_013308.1 |
1 - 1 |
2457 |
78 |
View Full Product Details