Assay Design
Gene Transcripts
- Gene Symbol
- IRC20
- Entrez Gene ID
-
850949
- Gene Name
- Gene Aliases
-
YLR247C
- Chromosome Location
-
Chr.XII: 628684 - 633354 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182134.1 |
NP_013348.1 |
1 - 1 |
4554 |
78 |
View Full Product Details