Assay Design
Gene Transcripts
- Gene Symbol
- NDL1
- Entrez Gene ID
-
850956
- Gene Name
- Gene Aliases
-
YLR254C
- Chromosome Location
-
Chr.XII: 644404 - 644973 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182141.1 |
NP_013355.1 |
1 - 1 |
448 |
97 |
View Full Product Details