Assay Design
Gene Transcripts
- Gene Symbol
- NKP2
- Entrez Gene ID
-
851025
- Gene Name
- Gene Aliases
-
YLR315W
- Chromosome Location
-
Chr.XII: 764808 - 765269 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182204.1 |
NP_013419.1 |
1 - 1 |
354 |
78 |
View Full Product Details