Assay Design
Gene Transcripts
- Gene Symbol
- SWC7
- Entrez Gene ID
-
851101
- Gene Name
- Gene Aliases
-
AWS1, YLR385C
- Chromosome Location
-
Chr.XII: 892992 - 893390 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182274.1 |
NP_013489.1 |
1 - 1 |
143 |
90 |
View Full Product Details