Assay Design
Gene Transcripts
- Gene Symbol
- VIP1
- Entrez Gene ID
-
851126
- Gene Name
- Gene Aliases
-
YLR410W
- Chromosome Location
-
Chr.XII: 937541 - 940981 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182298.1 |
NP_013514.1 |
1 - 1 |
3348 |
75 |
View Full Product Details