Assay Design



Gene Transcripts

Gene Symbol
ATG17
Entrez Gene ID
851142
Gene Name
Gene Aliases
APG17, YLR423C
Chromosome Location
Chr.XII: 971917 - 973170 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182311.3 NP_013527.3 1 - 1 1149 69

View Full Product Details