Assay Design
Gene Transcripts
- Gene Symbol
- SEC39
- Entrez Gene ID
-
851161
- Gene Name
- Gene Aliases
-
DSL3, YLR440C
- Chromosome Location
-
Chr.XII: 1015568 - 1017697 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182328.3 |
NP_013545.3 |
1 - 1 |
2051 |
65 |
View Full Product Details