Assay Design
Gene Transcripts
- Gene Symbol
- OST6
- Entrez Gene ID
-
854989
- Gene Name
- Gene Aliases
-
YML019W
- Chromosome Location
-
Chr.XIII: 233457 - 234455 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182377.1 |
NP_013693.1 |
1 - 1 |
846 |
101 |
View Full Product Details