Assay Design
Gene Transcripts
- Gene Symbol
- GAL80
- Entrez Gene ID
-
854954
- Gene Name
- Gene Aliases
-
YML051W
- Chromosome Location
-
Chr.XIII: 171594 - 172901 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182409.1 |
NP_013661.1 |
1 - 1 |
1245 |
86 |
View Full Product Details