Assay Design
Gene Transcripts
- Gene Symbol
- CUE4
- Entrez Gene ID
-
854871
- Gene Name
- Gene Aliases
-
YML101C
- Chromosome Location
-
Chr.XIII: 69735 - 70088 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182462.1 |
NP_013607.1 |
1 - 1 |
150 |
75 |
View Full Product Details