Assay Design
Gene Transcripts
- Gene Symbol
- PGA3
- Entrez Gene ID
-
854914
- Gene Name
- Gene Aliases
-
NQR1, YML125C
- Chromosome Location
-
Chr.XIII: 20761 - 21699 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182488.1 |
NP_013581.1 |
1 - 1 |
855 |
67 |
View Full Product Details