Assay Design
Gene Transcripts
- Gene Symbol
- BUD22
- Entrez Gene ID
-
855028
- Gene Name
- Gene Aliases
-
YMR014W
- Chromosome Location
-
Chr.XIII: 298868 - 300427 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182510.1 |
NP_013727.1 |
1 - 1 |
1344 |
92 |
View Full Product Details