Assay Design
Gene Transcripts
- Gene Symbol
- ARG7
- Entrez Gene ID
-
855084
- Gene Name
- Gene Aliases
-
ECM40, YMR062C
- Chromosome Location
-
Chr.XIII: 395054 - 396379 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182560.1 |
NP_013778.1 |
1 - 1 |
1157 |
86 |
View Full Product Details