Assay Design
Gene Transcripts
- Gene Symbol
- SAS2
- Entrez Gene ID
-
855157
- Gene Name
- Gene Aliases
-
KAT8, YMR127C
- Chromosome Location
-
Chr.XIII: 522329 - 523345 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182628.1 |
NP_013846.1 |
1 - 1 |
854 |
84 |
View Full Product Details