Assay Design
Gene Transcripts
- Gene Symbol
- MSS11
- Entrez Gene ID
-
855200
- Gene Name
- Gene Aliases
-
YMR164C
- Chromosome Location
-
Chr.XIII: 587274 - 589550 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182668.1 |
NP_013887.1 |
1 - 1 |
2148 |
92 |
View Full Product Details