Assay Design
Gene Transcripts
- Gene Symbol
- MMT1
- Entrez Gene ID
-
855215
- Gene Name
- Gene Aliases
-
MFT1, YMR177W
- Chromosome Location
-
Chr.XIII: 616566 - 618098 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182683.1 |
NP_013902.1 |
1 - 1 |
1450 |
84 |
View Full Product Details