Assay Design
Gene Transcripts
- Gene Symbol
- RGM1
- Entrez Gene ID
-
855220
- Gene Name
- Gene Aliases
-
YMR182C
- Chromosome Location
-
Chr.XIII: 624532 - 625167 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182688.1 |
NP_013907.1 |
1 - 1 |
554 |
77 |
View Full Product Details