Assay Design
Gene Transcripts
- Gene Symbol
- VTI1
- Entrez Gene ID
-
855237
- Gene Name
- Gene Aliases
-
YMR197C
- Chromosome Location
-
Chr.XIII: 658545 - 659198 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182704.1 |
NP_013924.1 |
1 - 1 |
449 |
105 |
View Full Product Details