Assay Design
Gene Transcripts
- Gene Symbol
- CLN1
- Entrez Gene ID
-
855239
- Gene Name
- Gene Aliases
-
YMR199W
- Chromosome Location
-
Chr.XIII: 662644 - 664284 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182706.1 |
NP_013926.1 |
1 - 1 |
1549 |
91 |
View Full Product Details