Assay Design
Gene Transcripts
- Gene Symbol
- PEP5
- Entrez Gene ID
-
855271
- Gene Name
- Gene Aliases
-
END1, VAM1, VPL9, VPS11, VPT11, YMR231W
- Chromosome Location
-
Chr.XIII: 733545 - 736634 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182738.1 |
NP_013958.1 |
1 - 1 |
2755 |
93 |
View Full Product Details