Assay Design
Gene Transcripts
- Gene Symbol
- SCS7
- Entrez Gene ID
-
855315
- Gene Name
- Gene Aliases
-
FAH1, YMR272C
- Chromosome Location
-
Chr.XIII: 809623 - 810777 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182779.1 |
NP_013999.1 |
1 - 1 |
951 |
71 |
View Full Product Details