Assay Design
Gene Transcripts
- Gene Symbol
- BUL1
- Entrez Gene ID
-
855318
- Gene Name
- Gene Aliases
-
DAG1, RDS1, SMM2, YMR275C
- Chromosome Location
-
Chr.XIII: 815651 - 818581 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182782.1 |
NP_014002.1 |
1 - 1 |
2646 |
75 |
View Full Product Details