Assay Design
Gene Transcripts
- Gene Symbol
- NIP1
- Entrez Gene ID
-
855357
- Gene Name
- Gene Aliases
-
YMR309C
- Chromosome Location
-
Chr.XIII: 892988 - 895426 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182820.1 |
NP_014040.1 |
1 - 1 |
2346 |
92 |
View Full Product Details