Assay Design
Gene Transcripts
- Gene Symbol
- ELP6
- Entrez Gene ID
-
855360
- Gene Name
- Gene Aliases
-
HAP3, KTI4, TOT6, YMR312W
- Chromosome Location
-
Chr.XIII: 898405 - 899226 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182823.1 |
NP_014043.1 |
1 - 1 |
749 |
80 |
View Full Product Details