Assay Design
Gene Transcripts
- Gene Symbol
- SUN4
- Entrez Gene ID
-
855659
- Gene Name
- Gene Aliases
-
SCW3, YNL066W
- Chromosome Location
-
Chr.XIV: 501515 - 502777 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182904.1 |
NP_014333.1 |
1 - 1 |
1145 |
103 |
View Full Product Details