Assay Design
Gene Transcripts
- Gene Symbol
- IBD2
- Entrez Gene ID
-
855557
- Gene Name
- Gene Aliases
-
YNL164C
- Chromosome Location
-
Chr.XIV: 325265 - 326320 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183002.1 |
NP_014235.1 |
1 - 1 |
945 |
105 |
View Full Product Details