Assay Design
Gene Transcripts
- Gene Symbol
- KEX2
- Entrez Gene ID
-
855483
- Gene Name
- Gene Aliases
-
QDS1, SRB1, VMA45, YNL238W
- Chromosome Location
-
Chr.XIV: 202428 - 204872 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183076.1 |
NP_014161.1 |
1 - 1 |
2351 |
100 |
View Full Product Details