Assay Design
Gene Transcripts
- Gene Symbol
- FOL1
- Entrez Gene ID
-
855465
- Gene Name
- Gene Aliases
-
YNL256W
- Chromosome Location
-
Chr.XIV: 164623 - 167097 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183094.1 |
NP_014143.2 |
1 - 1 |
2349 |
67 |
View Full Product Details