Assay Design
Gene Transcripts
- Gene Symbol
- PRM1
- Entrez Gene ID
-
855442
- Gene Name
- Gene Aliases
-
YNL279W
- Chromosome Location
-
Chr.XIV: 110917 - 112902 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183117.1 |
NP_014120.1 |
1 - 1 |
1853 |
95 |
View Full Product Details