Assay Design
Gene Transcripts
- Gene Symbol
- ATP11
- Entrez Gene ID
-
855401
- Gene Name
- Gene Aliases
-
YNL315C
- Chromosome Location
-
Chr.XIV: 43324 - 44280 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183153.1 |
NP_014084.1 |
1 - 1 |
851 |
90 |
View Full Product Details