Assay Design
Gene Transcripts
- Gene Symbol
- CSI2
- Entrez Gene ID
-
854155
- Gene Name
- Gene Aliases
-
YOL007C
- Chromosome Location
-
Chr.XV: 311342 - 312367 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183261.1 |
NP_014636.1 |
1 - 1 |
953 |
70 |
View Full Product Details