Assay Design
Gene Transcripts
- Gene Symbol
- HRD1
- Entrez Gene ID
-
854149
- Gene Name
- Gene Aliases
-
DER3, YOL013C
- Chromosome Location
-
Chr.XV: 301380 - 303035 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183267.1 |
NP_014630.1 |
1 - 1 |
1548 |
105 |
View Full Product Details