Assay Design
Gene Transcripts
- Gene Symbol
- NGL1
- Entrez Gene ID
-
854115
- Gene Name
- Gene Aliases
-
YOL042W
- Chromosome Location
-
Chr.XV: 249825 - 250916 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183296.1 |
NP_014600.1 |
1 - 1 |
746 |
104 |
View Full Product Details