Assay Design
Gene Transcripts
- Gene Symbol
- NBA1
- Entrez Gene ID
-
854084
- Gene Name
- Gene Aliases
-
YOL070C
- Chromosome Location
-
Chr.XV: 197220 - 198725 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183325.1 |
NP_014571.1 |
1 - 1 |
1450 |
85 |
View Full Product Details