Assay Design
Gene Transcripts
- Gene Symbol
- ATP19
- Entrez Gene ID
-
854077
- Gene Name
- Gene Aliases
-
YOL077W-A
- Chromosome Location
-
Chr.XV: 185438 - 185644 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184377.1 |
NP_014564.1 |
1 - 1 |
146 |
98 |
View Full Product Details