Assay Design



Gene Transcripts

Gene Symbol
HRP1
Entrez Gene ID
853997
Gene Name
Gene Aliases
NAB4, NAB5, YOL123W
Chromosome Location
Chr.XV: 87844 - 89448 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183377.1 NP_014518.1 1 - 1 1557 92

View Full Product Details