Assay Design
Gene Transcripts
- Gene Symbol
- AUS1
- Entrez Gene ID
-
854175
- Gene Name
- Gene Aliases
-
YOR011W
- Chromosome Location
-
Chr.XV: 349679 - 353863 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183430.1 |
NP_014654.1 |
1 - 1 |
4052 |
68 |
View Full Product Details